Skip to Content
Merck
All Photos(2)

Documents

Safety Information

MABC1216

Sigma-Aldrich

Anti-ARHGAP36 Antibody, clone 12D10.1

clone 12D10.1, from mouse

Synonym(s):

Rho GTPase-activating protein 36

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12D10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Rho GTPase-activating protein 36 (UniProt Q6ZRI8) is encoded by the ARHGAP36 gene (Gene ID 158763) in human. ARHGAP36 is a Rho GAP family member that acts as a potent hedgehog (Hh) pathway positive regulator. ARHGAP36 is reported to functionally associate with Hh pathway negative regulator Sufu (suppressor of fused) and ARHGAP36 overexpression results in reduced Gli3R levels, Gli2 ciliary accumulation, and Gli1 expression, in a Smo-independent manner. ARHGAP36 is produced with a signal peptide sequence (a.a. 1-40), which is removed posttranslationally to yield the mature protein with a Rho-GAP domain (a.a. 226-426). Unlike other Rho GAP domain-containing proteins, ARHGAP36 lacks the arginine finger motif that stimulates GTPase hydrolysis and GAP domain mutants can retain Gli-activating activity. ARHGAP36 isoforms are up-regulated in a subset of medulloblastomas, where they mediate noncanonical Gli activation by inhibiting Sufu.

Specificity

Clone 12D10.1 epitope lies within a C-terminal region present in all five human ARHGAP36 spliced isoforms reported by UniProt (Q6ZRI8).

Immunogen

GST-tagged recombinant human ARHGAP36 C-terminal fragment.

Application

Detect Arhgap36 using this mouse monoclonal Anti-ARHGAP36, clone 12D10.1 Antibody, Cat. No. MABC1216, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected ARHGAP36 in human prostate cancer, adrenal gland and pancreas tissue sections.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in Arhgap36 recombinant protein.

Western Blotting Analysis: A 1:500 dilution of this antibody detected 50 ng of GST-tagged recombinant human ARHGAP36 C-terminal fragment.

Target description

57.57/61.66 kDa (isoform 1 mature/proform), 57.57/58.34 kDa (isoform 2 mature/proform), 46.23 kDa (isoform 3), 57.57/60.57 kDa (isoform 4 mature/proform), 57.57 kDa (isoform 5).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MABC1216:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service