Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

MAB4410

Sigma-Aldrich

Anti-Renilla Luciferase Antibody, clone 1D5.2

ascites fluid, clone 1D5.2, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and Inventory is not available. Please check the Merck website for more information.

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1D5.2, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Recognizes Renilla luciferase.

Immunogen

Renilla luciferase GST fusion protein.

Application

Immunoblotting

EIA

Optimal working dilutions must be determined by end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-Renilla Luciferase Antibody, clone 1D5.2 is validated for use in ELISA, WB for the detection of Renilla Luciferase.

Target description

36 kDa

Physical form

Liquid.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Renilla

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MAB4410:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna E Mayrhofer et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(49), 31105-31113 (2020-11-25)
Kinase-targeted therapies have the potential to improve the survival of patients with cancer. However, the cancer-specific spectrum of kinase alterations exhibits distinct functional properties and requires mutation-oriented drug treatments. Besides post-translational modifications and diverse intermolecular interactions of kinases, it is
Ruth Röck et al.
Scientific reports, 5, 11133-11133 (2015-06-24)
Membrane receptor-sensed input signals affect and modulate intracellular protein-protein interactions (PPIs). Consequent changes occur to the compositions of protein complexes, protein localization and intermolecular binding affinities. Alterations of compartmentalized PPIs emanating from certain deregulated kinases are implicated in the manifestation
Hans H Schiffer et al.
Molecular pharmacology, 71(2), 508-518 (2006-09-14)
We have developed a new assay for measuring epidermal growth factor receptor (EGFR) activation using the bioluminescence resonance energy transfer (BRET) technology, which directly measures the recruitment of signaling proteins to activated EGFR. Our results demonstrate that EGFR BRET assays
Etienne Sauvageau et al.
Scientific reports, 7(1), 7148-7148 (2017-08-05)
AP-1 is a clathrin adaptor recruited to the trans-Golgi Network where it can interact with specific signals found in the cytosolic tail of cargo proteins to incorporate them into clathrin-coated vesicles for trafficking. The small G protein Arf1 regulates the
Graham J Belsham et al.
RNA (New York, N.Y.), 14(8), 1671-1680 (2008-06-24)
The initiation of protein synthesis on mRNAs within eukaryotic cells is achieved either by a 5' cap-dependent mechanism or through internal initiation directed by an internal ribosome entry site (IRES). Picornavirus IRES elements, located in the 5' untranslated region (5'UTR)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service