Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

MAB4356C3

Sigma-Aldrich

Anti-DPPA-2 Antibody, clone 6C1.2, Cy3 conjugate

clone 6C1.2, from mouse, CY3 conjugate

Synonym(s):

Pluripotent embryonic stem cell-related gene 1 protein, developmental pluripotency associated 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

CY3 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C1.2, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Dppa2(73703)

General description

DPPA-2 (developmental pluripotency associated-2) is a novel Oct-4-related protein that is localized to the nucleus of mouse fetal germ cells. DPPA-2 has binding sites for both POU and Sox-2 protein domains and is expressed primarily in the developing germ line following gastrulation. These associations suggest that this protein participates in a very strong supportive role in maintaining cell pluripotentiality. In addition to its role as a pluripotency marker, more recent research using cancer cells suggests that DPPA-2 may also act as an embryo-cancer antigen—pointing to the presence of abnormal cell growth and the formation of cancerous cells.

Immunogen

recombinant protein corresponding to mouse DPPA-2.

Application

Detect DPPA-2 using this Anti-DPPA-2 Antibody, clone 6C1.2, Cy3 conjugate validated for use in ICC.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Immunocytochemistry in Mouse Embryonic Stem Cells (SCR012). Immunocytochemsitry Analysis: A 1:100 dilution of this antibody detected DPPA2 in Mouse Embryonic Stem Cells (SCR012).

Target description

~30 kDa observed

Physical form

Protein G Purified
Purified mouse monoclonal IgG2aλ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8ºC protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Mouse Embryonic Stem Cells (SCR012).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MAB4356C3:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service