KE-1L-BK
Anti-K (cloneAEK4), FFMU
Anti-K from the clone AEK4, Human IgM, Intermediate for further manufacturing use
About This Item
clone
AEK4, monoclonal
Quality Level
form
liquid
packaging
pack of 1 L
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
General description
For the formulation of Anti-K blood typing reagents or devices.
Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Application
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
KE-1L-BK:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service