Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

HX0082

Supelco

Heptane, Biosynthesis

greener alternative

Synonym(s):

Heptane, n-Heptane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108

vapor pressure

48 hPa ( 20 °C)

Quality Level

Assay

≥99.0%
≥99.9% (saturated C7 hydrocarbons)

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm Water (H2O)

evapn. residue

≤1 ppm

color

APHA: ≤10

bp

97-98 °C/1013 hPa

mp

-90.5 °C

transition temp

flash point -4 °C

solubility

0.05 g/L

UV cutoff

≤200 nm

UV absorption

λ: 200 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.50
λ: 220 nm Amax: ≤0.20
λ: 250 nm Amax: ≤0.01
λ: 270 nm Amax: ≤0.005

greener alternative category

storage temp.

room temp

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent compared to hexane and thus align with "Safer Solvents and Auxiliaries". Click here for more information.

Application

Biosynthesis OMNISOLV®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (n-heptane):99.0% min
Assay (saturated C7 hydrocarbons):99.9% min
Color (APHA):10 max
Evaporation residue:1ppm max
Filtered through 0.2 μm filter:To pass test
Form:Clear liquid
Infrared Spectrum:Conforms to standard
UV Abs. at 200 nm:1.00AU max
UV Abs. at 210 nm:0.50AU max
UV Abs. at 220 nm:0.20AU max
UV Abs. at 250 nm:0.01AU max
UV Abs. at 270 nm:0.005AU max
UV Cut-off:200nm max
Water (H2O):100ppm max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

HX0082-1:
HX0082-901:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service