Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

HTS002M

Sigma-Aldrich

ChemiScreen CXCR2 Membrane Preparation

Human CXCR2 / IL-8b GPCR membrane preparation for Radioligand binding Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

CXCR2 is a 7-TM G-protein coupled receptor that binds to the chemokines GROalpha, GRObeta, GROgamma, IL-8, ENA-78, NAP-2 and GCP-2 (Olson and Ley, 2002). Neutrophils, mast cells and microvascular endothelial cells express CXCR2, and interactions of CXCR2 with its ligands promotes chemotaxis of these cell types (Heidemann et al., 2003; Nilsson et al., 1999; White et al., 1998). Studies with mice lacking CXCR2 indicate that CXCR2 promotes growth of primary tumors and secondary metastases (Keane et al., 2004), and plays an essential role in hyperoxia-induced lung injury (Sue et al., 2004). In addition, cytomegalovirus encodes a CXCR2-binding chemokine, vCXC-1, that promotes neutrophil migration to infected cells (Penfold et al., 1999). Chemicon′s CXCR2 Membrane Preps are ideal tools for screening for antagonists of interactions between CXCR2 and its ligands. Radiolabeled IL-8 binds to the CXCR2 membranes with a Kd of 0.075 nM. With 0.1 nM 125I-labeled IL-8, a 5-fold window of specific binding is obtained.
TRANSFECTION: Full-length human CXCR2 cDNA (Accession Number: M73969)

Application

Radioligand binding assay.

Biochem/physiol Actions

GPCR Class: A
Protein Target: CXCR2 / IL-8b
Target Sub-Family: Chemokine

Quality

Bmax for [125I]IL-8 binding: 0.93 pmol/mg protein; Kd for [125I] IL-8 binding: ~ 0.075 nM

Physical form

Membranes were adjusted to 0.5 mg protein/ml in 50 mM Tris pH 7.4, 10% glycerol and 1% BSA, rapidly frozen, and stored at -80°C. One vial typically contains 1000 units, where a unit is the amount of membrane that will yield greater than 1000 cpm specific binding of 125I-labeled IL-8 (0.1 nM input concentration).

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

HTS002M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service