FR180380074
Estapor® Magnetic Microspheres
MS-070/40 Mag PS 10%, Magnetic microspheres 1µm, ChemiLuminsecence Immunoassay
Synonym(s):
Magnetic Beads, Magnetic Particles
About This Item
Recommended Products
material
high-density polyethylene packaging (bottle)
polystyrene beads
Quality Level
composition
Iron Content, 35.0-50.0%
manufacturer/tradename
Estapor®
packaging
bottle of 10ml-10L
concentration
10% solids
technique(s)
enzyme immunoassay: suitable (ChemiLuminsecence Immunoassay-CLIA)
color
brown
matrix
polymeric polystyrene/divinylbenzene
bead diameter
0.70-1.30 μm
functional group
(Plain)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
FR180380074:
KR280380074:
FR180380074-100:
FR180380074-1000:
FR180380074-1:
FR180380074-VAR:
FR180380074-10:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service