Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

FR139482086

Sigma-Aldrich

K-030 Red Std PS 10%

Sign Into View Organizational & Contract Pricing


About This Item

NACRES:
NA.24
Pricing and availability is not currently available.

material

polystyrene

Quality Level

description

Functional Group Plain
<5% (Standard Deviation)

form

microspheres

concentration

10.0%

diam.

0.276-0.325 μm

color

red

Packaging

Polyethylene bottle

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

FR139482086-100:
FR139482086-1000:
FR139482086-VAR:
FR139482086-1:
FR139482086-10:
FR139482086:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service