Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

FCMAB172F

Sigma-Aldrich

Milli-Mark® Anti-CD5-FITC Antibody, clone DK23

clone DK23, Milli-Mark®, from mouse

Synonym(s):

T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DK23, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD5(921)

General description

CD5 is a transmembrane glycoprotein monomer with an Mr of 67,000. It possesses a large cytoplasmic domain suitable for signal transduction (Lozano, 1997). CD5 appears early in thymocyte development (Lozano, 1997), and is expressed at low density on thymocytes and at high density on all mature T lymphocytes (Lozano, 1997). The specificity of anti-CD5, DK23, is equivalent to that of the CD5-clustered antibody Leu-1 (Stein, 1984).

Specificity

Antibody recognizes Human CD5 antigen.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD5-FITC Antibody, clone DK23 is validated for use in FC for the detection of CD5.

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

67 kDa (Lozano, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C, protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
human lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

FCMAB172F:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service