Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

FCABS350F

Sigma-Aldrich

Milli-Mark® Anti-TrkA-FITC Antibody

Milli-Mark®, from rabbit

Synonym(s):

Neurotrophic tyrosine kinase receptor type, Oncogene TRK, TRK1-transforming tyrosine kinase protein, high affinity nerve growth factor receptor, neurotrophic tyrosine kinase, receptor, type 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

TrkA is a high affinity nerve growth factor (NGF). The Trk proto-oncogene family contains four members, TrkA, TrkB, TrkC, and TrkE, which are variably expressed throughout the central and peripheral nervous systems. TrkA binds to nerve growth factor (NGF) and autophosphorylates on tyrosine residues (Tyr490, Tyr674, Tyr675, Tyr751 and Tyr785) to activate multiple downstream effector proteins. Phosphorylation at Tyr490 is required for Shc association and subsequent activation of the Ras-MAP kinase-signaling cascade, which leads to activation of Elk-1-dependent gene transcription and neurite growth. Phosphorylations at Tyr674 and Tyr675 lie within the catalytic domain of TrkA tyrosine kinase and reflect Trk kinase activity. Additionally, phosphorylation at Tyr751 is required for PI3-kinase association and activation of the Akt signaling cascade.

Specificity

Antibody recognizes TrkA protein.
Unconjugated version Cat.No.06-574 is predicted to react with Rat based on immunogen design. The unconjugated part number was also shown to react with Mouse.

Immunogen

Bacterially expressed purified protein corresponding to the entire extracellular domain of rat TrkA receptor.
Epitope: Extracellular domain

Application

Detect TrkA using this Milli-Mark Anti-TrkA-FITC Antibody validated for use in FC.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

Evaluated by flow cytometry using HeLa cells.

Target description

TrkA exists as two isoforms: 87 kDa and 88 kDa

Physical form

Protein A purified
Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HeLa cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

FCABS350F:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service