Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

DR1123

Sigma-Aldrich

Anti-GCM1 Mouse mAb (2E11)

liquid, clone 2E11, Calbiochem®

Synonym(s):

Anti-Glial Cells Missing Homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E11, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCM1(8521)

General description

Anti-GCM1, mouse monoclonal, clone 2E11, recognizes the ~50 kDa GCM1 in FHs 173 WE and HeLa cells and human stomach tissue. It is validated for use in ELISA, WB, ICC, and IHC on paraffin sections.
Purified mouse monoclonal antibody. Recognizes the ~50 kDa GCM1 protein.
Recognizes the ~50 kDa GCM1 protein in FHs 173 WE and HeLa cells and human stomach tissue.

Immunogen

A recombinant polypeptide corresponding to amino acids of 108-166 human GCM1, expressed as a GST fusion protein

Application




ELISA (see comments)Immunoblotting (1-5 g/ml)Immunocytochemistry (10 g/ml)Paraffin Sections (1.7 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
FHs 173 WE cells, Human stomach tissue, HeLa cells

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

DR1123-100UG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service