CC1057
MMP-7, human, recombinant
Synonym(s):
Pro-matrilysin
About This Item
Recommended Products
biological source
human
Assay
~70% (followed by staining with CBB R-250, SDS-PAGE)
form
liquid
specific activity
5453 U/mg
manufacturer/tradename
Chemicon®
concentration
0.1 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
28,924 Daltons calculated from primary amino acid sequence.
Linkage
Unit Definition
Physical form
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
CC1057:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service