Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

BG-3

Sigma-Aldrich

X-Gal Stock Solution

The X-Gal is a histochemical substrate used to detect the presence of B-galactosidase.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.75
Pricing and Inventory is not available. Please check the Merck website for more information.

form

solution

manufacturer/tradename

Specialty Media

input

sample type: mouse embryonic stem cell(s)

General description

The X-Gal is a histochemical substrate used to detect the presence of B-galactosidase. It produces a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting and immunocytochemical assays. Cells expressing active B-galactosidase produce blue colonies when stained with X-Gal. It is one of the components required to visualize the expression of the LacZ reporter gene in cell culture.

Application

Research Category
All

Physical form

1ml liquid in DMSO at 40mg/ml.

Storage and Stability

Store at -20C upto 2 years

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

BG-3-G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service