Skip to Content
Merck
All Photos(1)

Documents

Safety Information

AGX819

Sigma-Aldrich

SARS-CoV-2 Spike Protein Recombinant Antigen

recombinant, expressed in HEK 293 cells

Synonym(s):

2019-nCoV, COVID-19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51201600
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

Quality Level

form

liquid

mol wt

≤138 kDa

storage condition

avoid repeated freeze/thaw cycles

shipped in

dry ice

General description

COVID-19 Spike Protein Recombinant Antigen, expressed in HEK293 cells, Cat. No. AGX819 is used as Positive Control.

Specificity

SARS-CoV-2

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 spike antigen can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).

Physical form

Purified protein in Tris Buffer with salts and additives without preservatives, pH 8.0.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

CR211549:
AGX819:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service