Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

ABT115

Sigma-Aldrich

Anti-FucT-III Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Galactoside 3(4)-L-Fucosyltransferase, Blood Group Lewis Alpha-4-Fucosyltransferase, Lewis Ft, Fucosyltransferase 3, Fucosyltransferase Iii, Fuct-Iii, Alpha-(1,3)-Fucosyltransferase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and Inventory is not available. Please check the Merck website for more information.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FUT3(2525)

General description

Alpha-(1,3)-fucosyltransferase 3 (FucT-III; Galactoside 3(4)-L-fucosyltransferase) is highly expressed in the small intestine, colon, stomach, and kidney. It is a member of the glycosyltransferase family of proteins which are involved in a glycosylation pathway that attaches fucose to Lewis epitopes of target molecules. This type of post-translational modification allows modified (fucosylated) molecules to recognize specific target substrates. Fucosyltransferases may therefore play an important role in communication at the cell surface. FucT-III may play a role in blood group determination as it is involved in the expression of antigens such as Lewis B, sialy Lewis X, Vim-2, and Lewis A.

Specificity

No homology to Rat and Mouse was found.
This antibody recognizes FucT-III at the lumenal region.

Immunogen

Epitope: Lumenal region
KLH-conjugated linear peptide corresponding to the lumenal region of human FucT-III.

Application

Anti-FucT-III Antibody detects level of FucT-III & has been published & validated for use in Western Blotting.
Research Category
Cell Structure
Research Sub Category
Glycobiology
Western Blot Analysis: 1 µg/mL from a representative lot detected FucT-III on 10 µg of transfected lysate and did not detect the non transfected lysate.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected FucT-III on 10 µg of human liver tissue lysate.

Target description

~50 kDa observed. The calculated molecular weight of this protein is 42 kDa, but can be observed at ~50 kDa due to high glycosylation. (Christensen, L, et al. (2000). Glycobiology. 10(9):931-939.)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human Liver Tissue Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ABT115:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service