Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

ABS2230

Sigma-Aldrich

Anti-USP46

new

Synonym(s):

Deubiquitinating enzyme 46, EC:3.4.19.12, Ubiquitin carboxyl-terminal hydrolase 46, Ubiquitin thioesterase 46, Ubiquitin-specific-processing protease 46

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 42.44 kDa
observed mol wt ~42 kDa

species reactivity

human

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 100

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... USP46(64854)

Specificity

This rabbit polyclonal antibody specifically detects USP46.

Immunogen

FLAG-tagged, full length recombinant protein of USP46.

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from USP46 (KO) 293T cells and wild type 293T cells.

Western Blotting Analysis (WB): A 1:1000 dilution of this antibody detected USP46 in lysate from wild type 293T cells, but not in USP46 (KO) 293T cell lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected USP46 in Western Blotting applications (Joo, et al. (2011) JBC 286(9); 7190-7201).

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected USP46 in Human kidney tissue sections.

Immunoprecipitation Analysis: A representative lot detected USP46 in Immunoprecipitation applications (Joo, et al. (2011) JBC 286(9); 7190-7201).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected USP46 in Chromatin Immunoprecipitation applications (Joo, et al. (2011) JBC 286(9); 7190-7201).

Immunocytochemistry Analysis: A representative lot detected USP46 in Immunocytochemistry applications (Huo Y. (2015) J. Neurochem. 134(6); 1067-1080).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Ubiquitin carboxyl-terminal hydrolase 46 (UniProt: P62068; also known as Deubiquitinating enzyme 46, Ubiquitin thioesterase 46, Ubiquitin-specific-processing protease 46, EC:3.4.19.12) is encoded by the USP46 gene (Gene ID: 64854) in human. USP46 gene encodes cysteine protease belonging to the ubiquitin-specific protease (USP) family of proteins. The protein contains a large USP domain (aa 35-365) and functions as a deubiquitinating enzyme capable of modifying cellular proteins. Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 cleaves ubiquitin from a ubiquitin-β-galactosidase fusion protein. These proteins regulate ubiquitin-mediated pathways that are involved in normal and pathological functions. It is also involved in the regulation of GABA action. Studies show that USP46, along with USP12, deubiquitinates both histone H2A and H2B. USP46 is predominantly localized in the cytoplasm. It is broadly expressed in multiple tissues, with high expression in heart muscle and CNS. Mutations or dysregulation of USP46 have been implicated in various diseases, particularly neuropsychiatric and neurological disorders. Additionally, USP46 may play a role in cancer, where its function in regulating protein stability and signaling pathways can contribute to tumor progression and resistance to therapy.(Ref.: Joo, et al. (2011). JBC 286(9); 7190-7201; Huo, Y., et al. (2015). J. Neurochem. 134(6); 1067-1080; Spencer, Z., et al. (2023). Nat Commun. 5;14(1); 6174).

Physical form

Rabbit polyclonal antiserum without azide.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ABS2230-100UL:
ABS2230-25UL:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service