Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

ABE1312

Sigma-Aldrich

Anti-SF3b155 (SAP 155) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Splicing factor 3B subunit 1, Pre-mRNA-splicing factor SF3b 155 kDa subunit, SF3b155, Spliceosome-associated protein 155, SAP 155

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SF3B1(23451)

General description

Splicing factor 3B subunit 1, also known as Pre-mRNA-splicing factor SF3b 155 kDa subunit (SF3b155), Spliceosome-associated protein 155 (SAP 155), and encoded by the gene name SF3B1 or SAP155, is a member of the SF3B1 family . SF3B155 (SAP 155) functions a s a subunit of the splicing factor SF3B required for ′A′ complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. SF3B155 (SAP 155) has been implied to be involved in the assembly of the ′E′ complex, and also may anchor U2 snRNP to the pre-mRNA. Additionally, SF3B155 (SAP 155) belongs to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron. SF3B155 (SAP 155) has been identified in the spliceosome C complex, as a component of the U11/U12 snRNPs that are part of the U12-type spliceosome. Phosphorylation of SF3B155 (SAP 155) occurs concomitantly with the splicing catalytic steps. The phosphorylation on Thr-244, Thr-248 and Thr-313 by cyclin-dependent kinases promotes interaction with PPP1R8 during mitosis. SF3B155 (SAP 155) is located in the nucleus speckle and during mitosis, it is transiently dispersed from the nuclear speckles to the cytoplasm.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human SF3b155 (SAP 155) near the N-terminus.

Application

Anti-SF3b155 (SAP 155) is an antibody against SF3b155 (SAP 155) for use in WB, ICC.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected SF3b155 (SAP 155) in HeLa, A431, and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected SF3b155 (SAP 155) in 10 µg of HeLa cell lysate.

Target description

~160 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ABE1312:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service