Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

ABC478

Sigma-Aldrich

Anti-phospho-CYLD Antibody (Ser418)

serum, from rabbit

Synonym(s):

Ubiquitin carboxyl-terminal hydrolase CYLD, Deubiquitinating enzyme CYLD, Ubiquitin thioesterase CYLD, Ubiquitin-specific-processing protease CYLD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer418)

Gene Information

human ... CYLD(1540)

General description

CYLD is a deubiquitinating enzyme that inactivates transcription factor NF-kappaB using TNFRs and is essential for normal cellular homeostasis of skin appendages. Deactivation or mutation of CYLD causes a loss of the deubiquitination activity and increases tumorigenesis. Loss of CYLD specifically results in familial cylindromatosis which is characterized by a predisposition to cylindromas, tumors of skin appendages. CYLD has been shown to cleave the lysine 63-linked polyubiquitin chain from proteins and the inactivity of CYLD has also been noticed in many other human cancers.

Specificity

This antibody detects phospho-CYLD at Ser418. The phosphorylation site of CYLD shares some sequence homology with serine phosphorylation site of c-Cbl.

Immunogen

Linear peptide corresponding to human phospho-CYLD (Ser418).

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-phospho-CYLD Antibody (Ser418) is validated for use in Western Blotting for the detection of phospho-CYLD (Ser418).
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-CYLD (Ser418) in 10 µg of human spleen and Jurkat lysate.
Western Blotting Analysis: A representative lot detected phospho-CYLD (Ser418) in PMA and Ionomycin-treated Jurkat cell lysate (Reiley, W., et al. (2005). Mole. Cell Biol. 25(10):3886-3895).

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-CYLD (Ser418) in 10 µg of Raji cell lysate.

Target description

~110 kDa observed.
Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ABC478:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service