Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

AB749P

Sigma-Aldrich

Anti-Collagen Type I Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
radioimmunoassay: suitable

suitability

not suitable for Western blot

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... COL1A1(1277)

Specificity

Reacts with bovine collagen type I collagen.

Antibody shows less than 0.1% cross reactivity in RIA at 1:500 dilution with bovine collagen types II, IX & XI, or fibronectin; other collagens and ECM proteins untested.

Reactivity with human collagen type I <2.0% with RIA, 1:500; reactivity with Chicken collagen type I, less than 0.1% by RIA at 1:500 dilution.

Immunogen

purified bovine type I collagen extracted from purified bovine skin.

Application

Anti-Collagen Type I Antibody detects level of Collagen Type I & has been published & validated for use in ELISA, RIA, IH, IH(P).
Immunohistochemistry: 1:40 dilution for immunofluorescent staining of frozen bovine liver and skin tissues.

Immunohistochemistry on 2% formalin or PFA fixed, paraffin embedded tissues requires HEIR antigen retrieval and enhanced enzymatic detection; methyl-carnoy, acetone, or acid-aldehyde fixation can enhance reactivity for paraffin sections.

Radioimmunoassay

ELISA: 1:200 direct

Not recommended for Western blots.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Linkage

Replaces: AB749

Physical form

Format: Purified
Protein G purified IgG fraction in liquid at 1 mg/mL in 0.01M phosphate, 0.09M NaCl, pH 7.2.No preservatives.

Storage and Stability

Maintain frozen at -20°C for up to 12 months. Avoid repeated freeze/thaw cycles

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB749P:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucio Díaz-Flores et al.
Scientific reports, 10(1), 4987-4987 (2020-03-21)
In lobular capillary hemangioma (LCH), misnamed pyogenic granuloma, only sprouting angiogenesis (SA) has been considered. We assess the occurrence of intussusceptive angiogenesis (IA) in LCH and whether IA determines the specific and other focal patterns in the lesion. For this
Lucio Díaz-Flores et al.
International journal of molecular sciences, 24(15) (2023-08-12)
Telocytes/CD34+ stromal cells (TCs/CD34+ SCs) have been studied in numerous organs and tissues, but their presence and characteristics in the parathyroid glands have not been explored. Using immunological and ultrastructural procedures, we assess the location, arrangement, and behavior of TCs/CD34+
Meng Wu et al.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada, 25(3), 822-829 (2019-04-27)
Tibetan mural samples from the Jiazhaer mountain cave were studied using enzyme-linked immunosorbent assays (ELISA) and immunofluorescence microscopy (IFM). Samples containing protein binders were first identified using ELISA, and then IFM was used to determine the location of protein binders.
Cemile Bektas et al.
Bioengineering (Basel, Switzerland), 11(8) (2024-08-31)
Cultivated meat, an advancement in cellular agriculture, holds promise in addressing environmental, ethical, and health challenges associated with traditional meat production. Utilizing tissue engineering principles, cultivated meat production employs biomaterials and technologies to create cell-based structures by introducing cells into
Lucio Díaz-Flores et al.
Histology and histopathology, 36(2), 217-228 (2020-12-19)
Intussusceptive angiogenesis (IA) is the process by which pre-existing blood vessels split, expand and remodel through intravascular pillar formation. In previous works, we studied the morphologic characteristics of intravascular papillary endothelial hyperplasia (IPEH) and suggested the participation of IA in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service