Skip to Content
Merck
All Photos(1)

Documents

Safety Information

AB6010

Sigma-Aldrich

Anti-MMP-12 Antibody, NT (active)

from rabbit, purified by affinity chromatography

Synonym(s):

Macrophage elastase, Matrix metalloproteinase-12, macrophage metalloelastase, matrix metallopeptidase 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP12(4321)

General description

Matrix metallopeptidase 12 (MMP-12) is able to degrade elastin, entactin, laminin-1, fibronectin, type IV collagen as well as insulin B-chain and casein. MMP-12 is structurally similar to the classical MMPs (MMP-1, MMP-3); it contains a propeptide with autoinhibitory cysteine switch site, a well conserved zinc site, hinge region and hemopexin domain. MMP-12 lacks a transmembrane domain and furin cleavage site. The zymogen for MMP-12 is about 54 kDa, and is quickly activated to the 45 kDa form, and this breaks down to cascade of active forms, ending with the common 22 kDa form.Stimulated macrophages produce MMP-12; it has also been found in osteosarcoma cells, synovial fibroblasts and lung fibroblasts. MMP-12 levels in quiescent cells and tissues are minimal (with the exception of macrophages), and mitogen stimulation or protein concentration is often needed to visualize the bands. In addition, cell types differ greatly in the quantity of MMP-12 produced.

Specificity

Sequence homology to mouse is 84%.
This antibody recognizes MMP-12 near the N-terminus of the active form.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to human MMP-12 near the N-terminus.

Application

Anti-MMP-12 Antibody, N-term (active) is an antibody against MMP-12 for use in WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Quality

Evaluated by Western Blot using a concentrated conditioned cell culture media collected from a proprietary human fibroblast cell line (Cat. No. AG902) and probing with 1 µg/mL of Anti-MMP-12.

Western Blot Analysis: 1 µg/ml of this antibody detected MMP-12 on 10 µg of lysate.

Target description

The zymogen for MMP-12 is ~ 54 kDa, and is quickly activated to a 45 kDa form. This breaks down to a cascade of active forms, ending with the common 22 kDa form.

Linkage

Replaces: AB19052

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Concentrated conditioned cell culture media collected from a proprietary human fibroblast cell line.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB6010:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandrine Bouchet et al.
Oncotarget, 7(15), 19445-19467 (2015-12-15)
The CD13 antigen's binding site for the Asn-Gly-Arg (NGR) motif enables NGR-containing chemotherapeutic drugs to be delivered to CD13-positive tumours. Human CD13-positive acute myeloid leukemia (AML) cells proliferate abnormally and escape death. Here, we show that the CNGRC-GG-D(KLAKLAK)2 peptide induces
Cayla Boycott et al.
Epigenetics, 17(11), 1513-1534 (2022-05-04)
Hepatocellular carcinoma (HCC) is mostly triggered by environmental and life-style factors and may involve epigenetic aberrations. However, a comprehensive documentation of the link between the dysregulated epigenome, transcriptome, and liver carcinogenesis is lacking. In the present study, Fischer-344 rats were
Yung-Wei Lin et al.
Cell death & disease, 11(7), 591-591 (2020-07-29)
N-α-Acetyltransferase 10 protein (Naa10p) was reported to be an oncoprotein in androgen-dependent prostate cancer (PCa; ADPC) through binding and increasing transcriptional activity of the androgen receptor (AR). PCa usually progresses from an androgen-dependent to an androgen-independent stage, leading to an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service