Skip to Content
Merck
All Photos(1)

Documents

Safety Information

AB3830

Sigma-Aldrich

Anti-Caveolin-1 Antibody, phospho-specific (Tyr14)

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr14)

Gene Information

human ... CAV1(857)

Specificity

Recognizes caveolin-1 only when phosphorylated at tyrosine 14. This antibody does not cross-react with caveolin-2, -3 or caveolin-1beta, or the short isoform of caveolin-1.

Immunogen

Epitope: phospho-specific (Tyr14)
KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Tyr14 of human caveolin-1

Application

This Anti-Caveolin-1 Antibody, phospho-specific (Tyr14) is validated for use in IP, WB for the detection of Caveolin-1.
Western blotting 1:1,000 (for best results, incubate membrane with diluted antibody in 5% BSA, 1X TBS, 0.1% Tween-20 at 4°C with gentle shaking, overnight)

Immunoprecipitation 1:50

Optimal working dilutions must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB3830:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service