Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

AB3611

Sigma-Aldrich

Anti-Survivin Antibody

Chemicon®, from rabbit

Synonym(s):

TIAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
mouse
technique(s):
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Apoptosis is triggered by a variety of stimuli, including members of the Tumor Necrosis Factor (TNF) family, and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family that binds to and inhibits cell death proteases. One of these IAP proteins, Survivin/TIAP, interacts with the processed form of caspase-3 and has been shown to inhibit its proteolytic activity. Survivin/TIAP is predominantly expressed in tissues of embryos, transformed cell lines, and many human cancers and lymphomas.

Specificity

Detects mouse survivin/TIAP in an approximate 17 kDa band by western blot.

Immunogen

Peptide corresponding to aa 128-140 of mouse survivin/TIAP.

Application

Anti-Survivin Antibody is an antibody against Survivin for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
Western blot: 1:500 to 1:1000

Mouse spleen tissue lysate can be used as a positive control.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at 4°C for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB3611:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yueyang Liu et al.
Oncology letters, 14(1), 525-532 (2017-07-12)
In the present study, the function of S100 calcium binding protein P (S100P) in the C666-1 nasopharyngeal carcinoma (NPC) cell line was examined. The levels of S100P protein in NPC tissues were analyzed using immunohistochemistry, and small interfering RNA silenced
Junghyun Kim et al.
Molecular medicine reports, 14(4), 3655-3661 (2016-09-08)
The aim of the present study was to assess the involvement of the high-mobility group box-1 (HMGB1) protein, receptor for advanced glycation end products (RAGE) and nuclear factor (NF)-κB signaling pathway in the development of diabetic retinopathy. Rat primary retinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service