Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

8850-OP

Millipore

TBE Buffer

OmniPur® Grade, 10X, Premixed Powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
Pricing and availability is not currently available.

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

color

white

shipped in

ambient

General description

Each pouch makes 1L of 10X TBE Buffer (1X = 0.089 M TRIS, 0.089M Borate, 0.002M EDTA). Molecular biology grade.

Specifications:
DNase: None detected
pH (1X concentration): at 25°C: 8.1-8.5

Protease: None detected
RNase: None detected
Solubility (1X concentration): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

8850-1PACK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service