Skip to Content
Merck
All Photos(1)

Key Documents

8.43943

Sigma-Aldrich

Propanephosphonic acid anhydride

(50% solution in 2-Methyltetrahydrofuran) for synthesis T3P

Synonym(s):

2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution, PPACA, Propylphosphonic anhydride, T3P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

bp

78-80 °C/1013 hPa (solvent)

transition temp

flash point -11 °C (solvent)

density

1.039 g/cm3 at 20 °C

storage temp.

2-8°C

Application


  • Study on the propylphosphonic anhydride (T3P) mediated Ugi-type three-component reaction. Efficient synthesis of an α-amino amide library: Explores the use of T3P in a three-component Ugi reaction, demonstrating its utility in synthesizing a diverse library of α-amino amides (Milen et al., 2017).

  • Cyclization of Linear Tetrapeptides Containing N‐Methylated Amino Acids by using 1‐Propanephosphonic Acid Anhydride: Reports the unexpected findings during the cyclization of linear tetrapeptides, where T3P played a crucial role in enhancing the reaction outcomes (Zhang et al., 2017).


Analysis Note

Assay (T3P) (NMR): ≥ 40.0 %
Pyropropanephosphonic acid (NMR): ≤ 10.0 %
Propanephosphonic acid (NMR): ≤ 0.50 %
2- Methyltetrahydrofurane (calculated): ≤ 50.0 %

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service