Skip to Content
Merck
All Photos(1)

Key Documents

8.20803

Sigma-Aldrich

3-Methylindole

for synthesis

Synonym(s):

3-Methylindole, Skatole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
201-471-7
Pricing and Inventory is not available. Please check the Merck website for more information.

Quality Level

Assay

≥98.5% (GC)

form

crystals

potency

3450 mg/kg LD50, oral (Rat)

bp

266 °C

mp

95-97 °C

transition temp

flash point 132 °C

storage temp.

2-30°C

InChI

1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3

InChI key

ZFRKQXVRDFCRJG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Melting range (lower value): ≥ 93 °C
Melting range (upper value): ≤ 97 °C
Identity (IR): passes test

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

269.6 °F

Flash Point(C)

132 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service