Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

8.14873

Sigma-Aldrich

Ethyl-2-amino-benzo(b)thiophene-3-carboxylate

for synthesis

Synonym(s):

Ethyl-2-amino-benzo(b)thiophene-3-carboxylate, 2-Amino-benzo[b]thiophene-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2S
CAS Number:
Molecular Weight:
221.28
UNSPSC Code:
12352108
Pricing and Inventory is not available. Please check the Merck website for more information.

Assay

≥96.0% (HPLC)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

bp

>290 °C/1013 hPa (decomposition)

mp

108.6 °C

storage temp.

2-30°C

InChI

1S/C11H11NO2S/c1-2-14-11(13)9-7-5-3-4-6-8(7)15-10(9)12/h3-6H,2,12H2,1H3

InChI key

XNASJEQIJMDBQN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 96.0 % (a/a)
Identity (IR): passes test

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

8148739999:
8.14873.0005A:
8148730000:
8148730001:
8148730005:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service