Skip to Content
Merck
All Photos(1)

Key Documents

8.14225

Sigma-Aldrich

Lithiumdiisopropylamide

(solution about 2 molar in cyclohexane/ethylbenzene/ tetrahydrofuran) for synthesis

Synonym(s):

Diisopropylaminolithium, LDA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
Pricing and availability is not currently available.

form

liquid

transition temp

flash point -25 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-8°C

General description

(solution about 2 molar in cyclohexane/ethylbenzene/ tetrahydrofuran) for synthesis

Analysis Note

Assay (acidimetric)26.0 - 34.0 %

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service