810663
Probumin® Bovine Serum Albumin Reagent Grade
≥98%, USA origin, 0.0-7.0 mg/dL Urea Nitrogen, enzyme immunoassay: suitable, Serology: suitable, Protein standard
About This Item
Recommended Products
product name
Probumin® Bovine Serum Albumin Reagent Grade,
biological source
bovine serum
Quality Level
type
for cell culture
Assay
≥98% (purity)
form
lyophilized powder
shelf life
48 mo.
technique(s)
enzyme immunoassay: suitable
protein quantification: suitable
dilution
( Serology)
foreign activity
Tested for IgG
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
810667:
81066N:
810664:
810662:
810661:
810663:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service