Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

8.00882

Sigma-Aldrich

Ethyl cyanoacetate

for synthesis

Synonym(s):

Ethyl cyanoacetate, Cyanoacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-309-0

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

460 °C

potency

>2000 mg/kg LD50, oral (Rat)

bp

209 °C/1013 hPa

mp

-22 °C

transition temp

flash point 110 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3

InChI key

ZIUSEGSNTOUIPT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.061 - 1.062
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

FSL

Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service