Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

6.09990

Supelco

Silver nitrate solution

c(AgNO3 )= 0.1 mol/ l (0.1N), concentrate for preparing 0.5 L volumetric solution for titration, Titrisol®

Synonym(s):

Silver nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302

Pricing and availability is not currently available.

Product Name

Silver nitrate solution, for 500 ml c(AgNO3 )= 0.1 mol/ l (0.1N) Titrisol

form

solution

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Antibacterial polymeric vesicles: A novel method for the in situ fabrication of silver nanoparticle-decorated polymeric vesicles was developed using silver nitrate, indicating significant antibacterial properties suitable for various biomedical applications (Zhang et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

IN26099900001:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service