Skip to Content
Merck
All Photos(1)

Documents

Safety Information

6002RU0

Sigma-Aldrich

Human Metapneumovirus IFA Reagent, RUO

Synonym(s):

hMPV IFA MPV, 6002RUO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

biological source

mouse

Quality Level

clone

monoclonal

manufacturer/tradename

Chemicon®

isotype

IgG

shipped in

dry ice

General description

Ready to use purified Anti-hMPV monoclonal antibodies in solution. Additional secondary FITC-labeled antibody may be used to visualize staining. The reagent is offered in a 2 mL dropper vial of monoclonal antibodies specific for Human Metapneumovirus (HMPV) in 0.2% BSA in PBS, Tween, with Sodium Azide. This is intended as a research use only and is not to be used for any other purpose.

Specificity

Human Metapneumovirus

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

2 mL dropper vial of monoclonal antibodies specific for Human Metapneumovirus (HMPV) in 0.2% BSA in PBS, Tween, with Sodium Azide.
Format: Purified

Storage and Stability

When stored at 2° to 8°C, reagent is stable up to the expiration date printed on the label.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

6002RU0:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prospective study of human metapneumovirus detection in clinical samples by use of light diagnostics direct immunofluorescence reagent and real-time PCR.
Landry, Marie L, et al.
Journal of Clinical Microbiology, 46, 1098-1100 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service