Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

573132-M

Millipore

STAT3 Inhibitor XVIII, BP-1-102

The STAT3 Inhibitor XVIII, BP-1-102 controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

STAT3 Inhibitor XVIII, BP-1-102, 4-(N-(4-Cyclohexylbenzyl)-2-(2,3,4,5,6-pentafluoro-N-methylphenylsulfonamido)acetamido)-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H27F5N2O6S
CAS Number:
Molecular Weight:
626.59
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Fc1c(c(c(c(c1F)F)[S](=O)(=O)N(CC(=O)N(Cc3ccc(cc3)C4CCCCC4)c2cc(c(cc2)C(=O)O)O)C)F)F

InChI

1S/C29H27F5N2O6S/c1-35(43(41,42)28-26(33)24(31)23(30)25(32)27(28)34)15-22(38)36(19-11-12-20(29(39)40)21(37)13-19)14-16-7-9-18(10-8-16)17-5-3-2-4-6-17/h7-13,17,37H,2-6,14-15H2,1H3,(H,39,40)

InChI key

WNVSFFVDMUSXSX-UHFFFAOYSA-N

General description

A cell-permeable, orally bioavailable amidosalicylic acid compound that selectively binds the three subpockets of Stat3 SH2 domain surface (KD = 504 nM) and blocks Stat3 phosphorylation, dimerization, and DNA-binding activity in a dose-dependent manner. Preferentially disrupts the high affinity binding between Stat3 and IL-6R/gp130-derived pTyr904 peptide (IC50 = 4.1 µM). Does not exert any significant effect on the phosphorylation status of Shc, Src, JAK1/2, Erk1/2, or Akt. Shown to reduce Stat3-dependent growth, survival, migration, and invasion of tumor cells in vitro and suppress the growth of aberrant Stat3-harboring human breast and lung tumor xenografts (3 mg/kg, i,v. or o.g.) in vivo.

Biochem/physiol Actions

Primary Target
Stat3

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, X., et al. Proc. Natl. Acad. Sci. USA109, 9623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

573132-10MG-M:
573132-10MG:
573132-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service