Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

559402

Sigma-Aldrich

SB 218078

A cell-permeable, ATP-competitive, and reversible indolocarbazole derivative that acts as a potent and selective inhibitor of checkpoint kinase (Chk1) in vitro. It inhibits Chk1 phosphorylation of cdc25C with an IC₅₀ of 15 nM.

Synonym(s):

SB 218078, 9,10,11,12,-Tetrahydro-9,12-epoxy-1 H-diindolo(1,2,3-fg:3ʹ,2ʹ,1ʹ-kl)pyrrolo(3,4-i)(1,6)benzodiazocine-1,3(2 H)-dione, Chk1 Inhibitor I, 9,10,11,12,-Tetrahydro-9,12-epoxy-1H-diindolo(1,2,3-fg:3ʹ,2ʹ,1ʹ-kl)pyrrolo(3,4-i)(1,6)benzodiazocine-1,3(2H)-dione, Chk1 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H15N3O3
CAS Number:
Molecular Weight:
393.39
UNSPSC Code:
12352200
Pricing and Inventory is not available. Please check the Merck website for more information.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C24H15N3O3/c28-23-19-17-11-5-1-3-7-13(11)26-15-9-10-16(30-15)27-14-8-4-2-6-12(14)18(22(27)21(17)26)20(19)24(29)25-23/h1-8,15-16H,9-10H2,(H,25,28,29)

InChI key

OTPNDVKVEAIXTI-UHFFFAOYSA-N

General description

A cell-permeable, ATP competitive, and reversible indolocarbazole derivative that acts as a potent inhibitor of checkpoint kinase (Chk1) in vitro (Ki = 15 nM). Binds to the ATP-binding pocket of Chk1, producing only slight changes in the protein conformation. Also inhibits the activity of CDK1 (Ki = 23 nM), CDK2 (Ki = 5.6 nM), and CDK4 (Ki = 16 nM). Inhibits Chk1 phosphorylation of Cdc25C (IC50 = 15 nM).
A cell-permeable, ATP-competitive, and reversible indolocarbazole derivative that acts as a potent and selective inhibitor of checkpoint kinase (Chk1) in vitro. It inhibits Chk1 phosphorylation of cdc25C with an IC50 of 15 nM. However, SB-218078 is a much weaker inhibitor of Cdc2 (IC50 = 250 nM) and PKC (IC50 = 1 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Chk1
Product competes with ATP.
Reversible: yes
Target IC50: 15 nM against Chk1 phosphorylation of cdc25C; 23 nM, 5.6 nM, 16 nM, against CDK1, CDK2, CDK4, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhao, B., et al. 2002. J. Biol. Chem.277, 46609.
Jackson, J.R., et al. 2000. Cancer Res.60, 566

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

559402-VMG:
559402-1MG:
559402-1.1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service