Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

553018-M

Millipore

RAF Kinase Inhibitor VII

Synonym(s):

RAF Kinase Inhibitor VII, 1-(4-(2,3-Dihydro-2-oxo-1H-imidazo[4,5-b]pyridin-7-yloxy)-2-(methylthio)phenyl)-3-(4-chloro-3-(trifluoromethyl)phenyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15ClF3N5O3S
CAS Number:
Molecular Weight:
509.89
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

caramel

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable imidazopyridinyl-urea compound that acts as a potent inhibitor against B-RafV600E activity (IC50 = 1 nM) by targeting its ATP-binding pocket and the adjacent lipophilic allosteric site accessible only when the enzyme is in its inactive DFG-out conformation. Reported to effectively inhibit RafV600E-dependent ERK phosphorylation (IC50 = 610 nM) and growth of human melanoma WM266.4 cells (GI50 = 470 nM).

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Menard, D., et al. 2009. J. Med. Chem.52, 3881.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

553018-5MG-M:
553018-MG:
553018-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service