Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

529313

Millipore

Polyoxin D, Streptomyces cacaoi var. asoensis

Specific inhibitor of fungal chitin synthetase (Ki = 6 µM).

Synonym(s):

Polyoxin D, Streptomyces cacaoi var. asoensis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N5O14
CAS Number:
Molecular Weight:
521.39
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

aqueous buffer: soluble
water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C17H23N5O14.Zn/c18-5(7(24)4(23)2-35-16(19)33)12(28)20-6(15(31)32)10-8(25)9(26)13(36-10)22-1-3(14(29)30)11(27)21-17(22)34;/h1,4-10,13,23-26H,2,18H2,(H2,19,33)(H,20,28)(H,29,30)(H,31,32)(H,21,27,34);/t4-,5-,6-,7+,8-,9+,10+,13+;/m0./s1

InChI key

ZKAPMXOKBGSLTC-AGVSZMOESA-N

General description

Specific inhibitor of fungal chitin synthetase (Ki = 6 µM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Acidic stock solutions are stable for up to 1 month at 20°C. Neutral or alkaline stock solutions are stable for up to 1 week at 20°C.

Other Notes

Causier, B.E., et al. 1994. Microbiol. 140, 2199.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

529313-1MG:
529313-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service