Skip to Content
Merck
All Photos(1)

Documents

Safety Information

528106

Sigma-Aldrich

PI 3-Kγ Inhibitor

The PI 3-Kγ Inhibitor, also referenced under CAS 648450-29-7, controls the biological activity of PI 3-Kγ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-Kγ Inhibitor, 5-Quinoxalin-6-ylmethylene-thiazolidine-2,4-dione, PI 3-K Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7N3O2S
CAS Number:
Molecular Weight:
257.27
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

potency

7.8 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pink

solubility

DMSO: 2.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C12H7N3O2S/c16-11-10(18-12(17)15-11)6-7-1-2-8-9(5-7)14-4-3-13-8/h1-6H,(H,15,16,17)/b10-6-

InChI key

SQWZFLMPDUSYGV-POHAHGRESA-N

General description

A cell-permeable thiazolidinedione compound that acts as a potent, selective, and ATP-competitive inhibitor of phosphatidylinositol 3-kinase γ (PI 3-Kγ) (Ki = 7.8 nM; IC50 = 8 nM, 60 nM, 270 nM, 300 nM for p110-γ, α, β and δ-isoforms, respectively). Does not affect the activity of a wide panel of kinases, receptors, enzymes, and ion channels even at concentrations as high as 1 µM. Selectively blocks MCP-1-, but not CSF-1-, dependent Akt phosphorylation and chemotaxis in primary Pik3cg+/+ murine monocytes and in a human monocytic cell line THP-1 (IC50 = 181 nM). Also shown to exhibit in vivo efficacy in reducing neutrophil chemotaxis in murine peritonitis and arthritis models.
A cell-permeable, potent, selective, and ATP-competitive inhibitor of phosphatidylinositol 3-kinase γ (PI 3-Kγ) (Ki = 7.8 nM; IC50 = 8 nM, 60 nM, 270 nM, 300 nM for p110-γ, α, β and δ-isoforms, respectively). Does not affect the activity of a wide panel of kinases, receptors, enzymes, and ion channels even at concentrations as high as 1 µM. Selectively blocks MCP-1-, but not CSF-1-, dependent Akt phosphorylation and chemotaxis in primary Pik3cg+/+ murine monocytes and in a human monocytic cell line THP-1 (IC50 = 181 nM). Also shown to exhibit in vivo efficacy in reducing neutrophil chemotaxis in murine peritonitis and arthritis models.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Phosphatidylinositol 3-kinase γ (PI 3-Kγ)
Product competes with ATP.
Reversible: no
Target IC50: 8 nM, 60 nM, 270 nM, 300 nM for p110-γ, α, β and δ-isoforms, respectively; 181 nM in selectively blocking MCP-1-, but not CSF-1-, dependent Akt phosphorylation and chemotaxis in primary Pik3cg+/+ murine monocytes and in a human mono

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Barber, D.F., et al. 2005. Nat. Med.11, 933.
Camps, M., et al. 2005. Nat. Med.11, 936.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

528106-VMG:
528106-5MG:
528106-500MG:
528106-1.1ML:
528106-250MG:
528106-100MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service