Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

525146-M

Millipore

sPLA2-IIA Inhibitor II, KH064

Synonym(s):

(S)-5-(4-(benzyloxy)phenyl)-4-(7-phenylheptanamido)pentanoic acid, Phospholipase A₂ Group IIA Inhibitor II, PLA2g2a Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A reversible, potent, and selective sPLA2-IIA/PLA2g2a (phospholipase A2 group IIA) inhibitor (IC50 = 29 nM) that displays much reduced potency against sPLA2-V/PLA2g5 (PLA2 group V; IC50 = 2 µM) and is reported not to affect the activity of cytosolic cPLA2, Ca2+-independent iPLA2, 30 human GPCRs, and 25 other enzymes. In addition to being useful in studying PLA2g2a-mediated cellular responses in cultures in vitro (100% inhibition of LPS-induced PGE2 production in HMC-1 cells; 15 min 10 µM pretreatment), KH064 is orally available in rats and efficacious in various rat disease models in vivo (5 mg/kg p.o.), including adjuvant-induced arthritis, spontaneous hypertension, high fat diet-induced metabolic disorders. Comparing to the substrate-competitive inhibitor c(2NapA)LS(2NapA)R (Cat. No. 525145), KH064 also effectively chelates sPLA2-IIA active site Ca2+ in addition to targeting the enzyme′s N-terminal lipid substrate binding site.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hansford, K. A., et al. 2003. Chembiochem.4, 181.
Iyer, A., et al. 2012. Diabetes.61, 2320,
Levick, S., et al. 2006. J Immunol.176, 7000.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

525146-10MG:
525146-10MG-M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service