Skip to Content
Merck
All Photos(1)

Documents

5.09892

Sigma-Aldrich

Tetrabenazine, TBZ

Synonym(s):

Tetrabenazine, TBZ, VMAT2 Inhibitor, Xenazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

InChI

1S/C19H27NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16H,5-7,10-11H2,1-4H3

InChI key

MKJIEFSOBYUXJB-UHFFFAOYSA-N

General description

A cell permeable benzoquinolizine based compound that acts as a monoamine-depleting agent by blocking the activity vesicular monoamine transporter 2. Promotes late-stage differentiation of Pdx1-positive pancreatic progenitor cells into Neurog3 (Ngn3)-positive endocrine precursors without increasing their proliferation. Increases the number of insulin expressing cells in a dose dependent manner (EC50 = 220 nM) and this effect is significantly enhanced when cells are simultaneously treated with dibutyryl cAMP. Embryonic stem cells treated with TBZ and/or dibutyryl cAMP and then grafted into kidney capsule of AKITA mice reduce hyperglycemia, improve fasting blood glucose levels, and show an increase in plasma C-peptide levels.
A cell permeable benzoquinolizine based compound that acts as a monoamine-depleting agent by blocking the activity vesicular monoamine transporter 2. Promotes late-stage differentiation of Pdx1-positive pancreatic progenitor cells into Neurog3 (Ngn3)-positive endocrine precursors without increasing their proliferation. Increases the number of insulin expressing cells in a dose dependent manner (EC50 = 220 nM) and this effect is significantly enhanced when cells are simultaneously treated with dibutyryl cAMP. Embryonic stem cells treated with TBZ and/or dibutyryl cAMP and then grafted into kidney capsule of AKITA mice reduce hyperglycemia, improve fasting blood glucose levels, and show an increase in plasma C-peptide levels.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
VMAT2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sakano, D., et al. 2014. Nat. Chem. Bio.10,141.

Jankovic, J., et al. 1997. Neurobiology48,1358.
Gertrude, P.Q., et al. 1959. J. Pharm. Exp. Ther.127, 103.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service