Skip to Content
Merck
All Photos(1)

Key Documents

5.09290

Millipore

G 418 Sulfate Salt

OmniPur® Grade, >=730 µg/mg

Synonym(s):

OmniPur® G418 Sulfate, Cell Culture Tested, NMD Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

cation traces

heavy metals: ≤100 ppm

storage temp.

10-30°C

InChI

1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1

InChI key

UHEPSJJJMTWUCP-NKCAIAFTSA-N

General description

Aminoglycoside antibiotic related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis, is toxic to bacteria, yeast, protozoans, helminths, higher plant and mammalian cells. Used in molecular genetics as a selective agent for the bacterial neor/kanr genes. The product of these genes, aminoglycoside 3′-phosphotransferase, inactivates G418, neomycin, and kanamycin by phosphorylation. Introduction of either of these genes into cells can confer resistance to G418, which enables cells to grow in media containing G418.
Aminoglycoside related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis. Toxic to bacteria, yeast, protozoans, helminths, higher plant, and mammalian cells. Widely used in the selection of eukaryotic expression vectors carrying the bacterial neor/kanr genes. The product of these genes, aminoglycoside 3′-phosphotransferase, inactivates G418, neomycin, and kanamycin by phosphorylation. Introduction of either of these genes into cells can confer resistance to G418, which enables cells to grow in media containing G418.

Biochem/physiol Actions

Assay: ≥730 µg/mg

Warning

Toxicity: Standard Handling (A)

Reconstitution

Solubilization in H₂O at 100 mg/ml yields a clear, colorless solution.

Other Notes

Ethier, S.P., and Taback, E. 1993. Cancer Lett.74, 189.
Santerre, R.F., et al. 1991. Methods Mol. Biol.7, 245.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition, Cold Spring Harbor, NY.
Edwards, S.A., and Adamson, E.D. 1987. J. Cell Physiol.133, 46.
Ernst, J.F., and Chan, R.K. 1985. J. Bacteriol.163, 8.
Canaani, D., and Berg, P. 1982. Proc. Natl. Acad. Sci. USA79, 5166.
Hirth, K.P., et al. 1982. Proc. Natl. Acad. Sci. USA79, 7356.
Ursic, D., et al. 1981. Biochem. Biophys. Res. Commun.101, 1031.
Jimenez, A., and Davies, J. 1980. Nature287, 869.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service