Skip to Content
Merck
All Photos(1)

Key Documents

5.00713

Sigma-Aldrich

EphA Antagonist

Synonym(s):

EphA Antagonist, Rtk2 Antagonist, Rtk1 Antagonist, 4-(2,5-dimethyl-1H-pyrrol-1-yl)-2-hydroxybenzoic acid, EphA2 Antagonist, EphA4 Antagoinst, Ephrin Receptor Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO3
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

3 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A pyrrolyl benzoic acid that effectively competes against ephrin-B3 for EphA4 binding as well as against ephrin-A1/-A2/-A3/-A5 for EphA2 and EphA4 binding by targeting EphA2/A4 high affinity ephrin-binding pocket in a reversible manner (Ki = 6-10 and 11-14 µM against ephrin-A5 binding to EphA4 and EphA2, respectively), exhibiting much reduced or no potency against ephrin-A4 for EphA2/EphA4 binding, ephrin-A5 for EphA3/A5/A6/A7 binding, ephrin-B2 for B1/B2/B3/B4/B6 binding, or ephrin-B2 for EphA4 binding. Shown to inhibit 0.1 µg/mL ephrin-A1-Fc fusion-induced EphA2 phosphorylation in COS7 (by 92% at 200 µM) and 0.5 µg/mL ephrin-A5-Fc fusion-induced EphA4 phosphorylation in HT22 neuronal cultures (by 42% and 79% respectively, at 300 and 400 µM) without any cytotoxicity (up to 400 µM and 3 days in HT22 cultures). Also effective toward chicken and zebrafish EphA4 orthologs as evidenced by its efficacy in preventing 1 µg/mL ephrin-A5-Fc-induced growth cone collapse of neurons in 6-d chicken embryo retina (by 100% at 400 µM) ex vivo and in fully rescuing human mutant SOD-1 A4V transgene-induced axonopathy in zebrafish embryos (500 µM; treatment period from 6 to 30 hpf) in vivo.
A pyrrolyl benzoic acid that effectively competes against ephrin-B3 for EphA4 binding as well as against ephrin-A1/-A2/-A3/-A5 for EphA2 and EphA4 binding by targeting EphA2/A4 high affinity ephrin-binding pocket in a reversible manner, exhibiting much reduced or no potency against ephrin-A4 for EphA2/EphA4 binding, ephrin-A5 for EphA3/A5/A6/A7 binding, ephrin-B2 for B1/B2/B3/B4/B6 binding, or ephrin-B2 for EphA4 binding. Shown to inhibit ephrin-A1-induced EphA2 phosphorylation (by 92% at 200 µM) and ephrin-A5-induced EphA4 phosphorylation (by 79% at 400 µM) in cultures in vitro and prevent ephrin-A5-induced growth cone collapse of neurons in 6-d chicken embryo retina (by 100% at 400 µM) ex vivo, as well as rescue human mutant SOD-1 A4V transgene-induced axonopathy in zebrafish embryos (500 µM from 6 to 30 hpf) in vivo.

Biochem/physiol Actions

Cell permeable: no
Primary Target
EphA
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Van Hoecke, A., et al. 2012. Nat. Med.18, 1418.
Noberini, R., et al. 2008. J. Biol. Chem.283, 29461.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service