Skip to Content
Merck
All Photos(1)

Documents

Safety Information

475878

Sigma-Aldrich

(+)-MK 801 Maleate

A highly potent, selective, and non-competitive NMDA receptor antagonist that acts by binding to a site located within the NMDA-associated ion channel, thus preventing Ca2+ flux.

Synonym(s):

(+)-MK 801 Maleate, (5R,10S)-(+)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d]-cyclohepten-5-10-imine, Maleate, Dizocilpine Maleate, NMDA Antagonist I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N · xC4H4O4
CAS Number:
Molecular Weight:
221.30 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 3 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H15N.C4H4O4/c1-16-13-8-4-2-6-11(13)10-15(17-16)12-7-3-5-9-14(12)16;5-3(6)1-2-4(7)8/h2-9,15,17H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-,16+;/m1./s1

InChI key

QLTXKCWMEZIHBJ-PJGJYSAQSA-N

General description

A highly potent, selective and non-competitive NMDA receptor antagonist that acts by binding to a site located within the NMDA associated ion channel, preventing Ca2+ flux. An effective anti-ischemic agent in several animal models. Has been used to develop an in vivo model of NMDA receptor-dependent apoptotic neurodegenerative disease in developing rat brains. Has also been used to develop an in vitro model of NMDA receptor-specific Ca2+ entry induced seizures.
A highly potent, selective, and non-competitive NMDA receptor antagonist that acts by binding to a site located within the NMDA-associated ion channel, thus preventing Ca2+ flux. An effective anti-ischemic agent in several animal models. Has been used to develop an in vivo model of NMDA receptor-dependent apoptotic neurodegenerative disease in developing rat brains. Has also been used to develop an in vitro model of NMDA receptor-specific Ca2+ entry induced seizures.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NMDA receptor
Reversible: no

Warning

Toxicity: Standard Handling (A)

Preparation Note

Stock solutions in water may require warming and vigorous stirring to achieve complete solubilization.

Reconstitution

Following reconstitution in water refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Aqueous stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.

Other Notes

Ikonomidou, C., et al. 1999. Science 283, 70.
DeLorenzo, R.J., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 14482.
Woodruff, G.N., et al. 1987. Neuropharmacology 26, 903.
Wong, E.H.F., et al. 1986. Proc. Natl. Acad. Sci. USA 83, 7104.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

475878-10MG:
475878-MG:
475878-1.1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service