475741
MAGL Inhibitor III, JZL184
The MAGL Inhibitor III, JZL184, also referenced under CAS 1101854-58-3, controls the biological activity of MAGL.
Synonym(s):
MAGL Inhibitor III, JZL184, 4-Nitrophenyl-4-(dibenzo[d][1,3]dioxol-5-yl(hydroxy)methyl)piperidine-1-carboxylate, Monoacylglycerol Lipase Inhibitor III, JZL184
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
DMSO: 50 mg/mL
shipped in
wet ice
storage temp.
−20°C
InChI
1S/C27H24N2O9/c30-26(38-21-5-3-20(4-6-21)29(32)33)28-11-9-17(10-12-28)27(31,18-1-7-22-24(13-18)36-15-34-22)19-2-8-23-25(14-19)37-16-35-23/h1-8,13-14,17,31H,9-12,15-16H2
InChI key
SEGYOKHGGFKMCX-UHFFFAOYSA-N
General description
Warning
Reconstitution
Other Notes
Long, J.Z., et al. Nature Chem. Biol.16, 744.
Long, J.Z., et al. 2009. Nature Chem. Biol.5, 37.
Pan, B., et al. 2009. J. Pharmacol. Exp. Ther.331, 591.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
475741-10MG:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service