Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

454863

Sigma-Aldrich

D-erythro-MAPP

A synthetic analog of ceramide.

Synonym(s):

D-erythro-MAPP, (1S,2R)-D- erythro-2-(N-Myristoylamino)-1-phenyl-1-propanol, AC Inhibitor II, Acid Ceramidase Inhibitor II, D-e-MAPP, (1S,2R)-D-erythro-2-(N-Myristoylamino)-1-phenyl-1-propanol, AC Inhibitor II, Acid Ceramidase Inhibitor II, D-e-MAPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H39NO2
CAS Number:
Molecular Weight:
361.56
UNSPSC Code:
12352211

Quality Level

Assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C23H39NO2/c1-3-4-5-6-7-8-9-10-11-12-16-19-22(25)24-20(2)23(26)21-17-14-13-15-18-21/h13-15,17-18,20,23,26H,3-12,16,19H2,1-2H3,(H,24,25)/t20-,23-/m1/s1

InChI key

YLAZEWZHIRBZDA-NFBKMPQASA-N

General description

A synthetic analog of ceramide. Shown to selectively inhibit alkaline ceraminidase both in vitro and in HL-60 cells (IC50 = 1-5 µM for alkaline ceramides versus IC50 >500 µM for acid ceraminidase). This inhibition results in and over 3-fold increase in endogenous ceramide in treated cells due to a lack of breakdown of ceramide to sphingosine.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Alkaline ceramidase
Product does not compete with ATP.
Reversible: no
Target IC50: 1-5 µM for alkaline ceramidase versus >500 µM for acid ceramidase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Bielawska, A., et al. 1996. J. Biol. Chem. 271, 12646.
Hannun, Y.A. 1996. Science274, 1855.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

454863-MG:
454863-1MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service