Skip to Content
Merck
All Photos(1)

Documents

Safety Information

420220-M

Millipore

Thiazovivin

≥98% (HPLC), powder, Rho kinase inhibitor, Calbiochem

Synonym(s):

iPSC Induction Enhancer, Thiazovivin, N-Benzyl-2-(pyrimidin-4-ylamino)thiazole-4-carboxamide, Induced Pluripotent Stem Cells Induction Enhancer, Rho Kinase Inhibitor XII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N5OS
Molecular Weight:
311.36
UNSPSC Code:
12352200

product name

iPSC Induction Enhancer, Thiazovivin, A cell-permeable aminothiazolo-carboxamide compound that is reported to augment SB431542 and PD0325901.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable aminothiazolo-carboxamide compound that is ~5-fold more potent than Y-27632 (Cat. Nos. 688000 & 688001) in inhibiting ROCK enzymatic activity (71% inhibition at 2 µM). Single-cell dissociation of cultured hESCs is demonstrated to hyperactivate Rho-ROCK signaling and result in cell death, while suppression of such hyperactivation by Tzv (2 µM) or Y-27632 (2 µM) stabilize newly synthesized cell surface E-cadherin, which in turn promotes cell-cell adhesion and prevents trypsinization-induced apoptosis (12% vs 81% TUNEL-positive cells after trypsinization in matrigel cultures with 2 µM Tzv or DMSO, respectively). Unlike, Ptn (Cat. No. 544049), Tzv protects dissociated individual hESCs even in the absence of ECM (24% vs 90% TUNEL-positive cells after trypsinization in non-coating plates with 2 µM Tzv or Ptn, respectively). Simultaneous blockage of TGFβ and MEK-ERK pathways by 2 µM SB431542 (Cat. No. 616461)and 0.5 µM PD0325901 (Cat. No. 444966) is reported to result in >50-fold improvement of human iPSCs (induced Pluripotent Stem Cells) generation from fibroblast cultures virally transfected with 4-TF (Oct44, Klf4, Sox2, and c-Myc), while the inclusion of 0.5 µM Tzv on top of SB431542 and PD0325901 is demonstrated to further improve the efficiency to an overall improvement of >200-fold from 4-TF-induction alone.
A cell-permeable aminothiazolo-carboxamide compound that is ~5-fold more potent than Y-27632 (Cat. Nos. 688000 & 688001) as a ROCK inhibitor (71% inhibition at 2 µM). Effectively prevents trypsinization-induced apoptosis in hESC cultures by stabilizing newly synthesized cell surface E-cadherin and cell-cell adhesion. Unlike, Ptn (Cat. No. 544049), Tzv protects dissociated hESCs even in the absence of ECM. Greatly augments SB431542 (Cat. No. 616461)and PD0325901 (Cat. No. 444966) in improving iPSCs generation efficiency from 4-TF- (Oct44, Klf4, Sox2, and c-Myc) transfected fibroblast cultures.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xu, Y., et al. 2010. Proc. Natl. Acad. Sci. USA107, 8129
Lin, T., et al. 2009. Nat. Methods6, 805.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

420220-10MG:
420220-MG:
420220-10MG-M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service