Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

401490

Sigma-Aldrich

IKK-2 Inhibitor XI

The IKK-2 Inhibitor XI, also referenced under CAS 354810-80-3, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IKK-2 Inhibitor XI, (5-Phenyl-3-ureido)-thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N3O2S
CAS Number:
Molecular Weight:
261.30
UNSPSC Code:
12352200
Pricing and Inventory is not available. Please check the Merck website for more information.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thiophenecarboxamide-ureido compound that acts as a potent, ATP-binding pocket-targeting inhibitor of IKK-2 (IC50 = 25 nM), while affecting IKK-1 and JNK1 only at much higher concentrations (IC50 = 1.0 and 1.6 µM, respectively). Shown to reduce LPS-induced TNF-α production in PBMCs (IC50 = 250 nM) and exhibit oral availability with good pharmacokinetic in rats in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Baxter, A., et al. 2004. Bioorg. Med. Chem. Lett.14, 2817.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

401490-5MG:
401490-1.1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service