Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

382181

Sigma-Aldrich

HDAC Inhibitor XXIV, OSU-HDAC-44

Synonym(s):

HDAC Inhibitor XXIV, OSU-HDAC-44, OSU-HDAC-44, 4-(2,2-Dimethyl-4-phenylbutanamido)-N-hydroxybenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O3
CAS Number:
Molecular Weight:
326.39
UNSPSC Code:
12352200
Pricing and Inventory is not available. Please check the Merck website for more information.

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, phenylbutyrate hydroxamate pan-HDAC inhibitor that is shown to suppress the activities of class I (HDAC1 and HDAC8), class II (HDAC4 and HDAC6), and class IV (HDAC11) HDACs by > ~90% compared to control, at a concentration of 1 µM with a comparable or greater inhibitory effect than SAHA. In vitro, this compound exhibits a broad spectrum of anti-tumor activities in NSCLC cells, holds synergistic effects with cisplatin in the reduction of cell viability, is implicated in the induction of the intrinsic apoptotic pathway, and displays inhibitory effects against mitosis and cytokinesis. This inhibitor is also shown to suppress tumor growth in mice genograft models by 62%, 78% and 90% at 7.5, 15 and 30 mg/kg respectively without adversely affect in body weight and detectable toxicity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tang, Y., et al. 2010. PLoS One5, e12417.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

382181-10MG:
382181-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service