Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

347425

Millipore

Gluconolactone

An oxidation product of glucose by glucose oxidase.

Synonym(s):

Gluconolactone, Glucono-δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

Assay

≥99% (acidimetric)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2

InChI key

PHOQVHQSTUBQQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A ketoaldehyde resulting from the oxidation of glucose by glucose oxidase. Acts as a mixed type inhibitor of glucoamylase, possibly by forming a nonproductive ternary complex with the enzyme and the substrate.
An oxidation product of glucose by glucose oxidase.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Tanaka, A., et al. 1999. Biosci. Biotechnol. Biochem.63, 1548.
Rahbar, S., and Nadler, J.L. 1999. Clin. Chim. Acta287, 123.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

347425-GM:
347425-500GM:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service