Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

344045

Sigma-Aldrich

FOCUS-Protease Arrest Cocktail

A protease cocktail specifically developed for 2D gel sample preparation.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

2-8°C

General description

A protease cocktail specifically developed for 2D gel sample preparation.
A protease inhibitor cocktail specifically developed for the inhibition of protease activity during protein purification and sample processing. Suitable for use with yeast, bacterial, plants, and mammalian tissues. No EDTA is used in this preparation. Compatible with FOCUS-Nuclease for removing nucleic acid contamination from samples. FOCUS-Protease-Arrest contains agents that inhibit serine, cysteine, metallo-proteases as well as calpains. FOCUS-Protease-Arrest contains both irreversible and reversible protease inhibitors. FOCUS-Protease Arrest, used at 1:50 dilution in extraction buffer (pH 7-8) inhibits over 95-98% of protease activities.

Biochem/physiol Actions

Primary Target
Serine, cysteine, metalloproteinases, and calpain proteases
Product does not compete with ATP.

Warning

Toxicity: Irritant (B)

Physical form

Supplied as 100X.

Reconstitution

Following dilution in extraction/lysis buffer or extract, the FOCUS-Protease Arrest inhibitors are stable for up to 2 weeks at 4°C or up to 4 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

344045-ML:
344045-1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service