Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

324896

Sigma-Aldrich

eIF-2α Inhibitor II, Sal003

The eIF-2α Inhibitor II, Sal003, also referenced under CAS 301359-91-1, controls the biological activity of eIF-2α. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

eIF-2α Inhibitor II, Sal003, 3-Phenyl-N-(2,2,2-trichloro-1-((((4-chlorophenyl)amino)carbonothioyl)amino)ethyl)acrylamide, Sal003

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15Cl4N3OS
CAS Number:
Molecular Weight:
463.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and Inventory is not available. Please check the Merck website for more information.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable Salubrinal (Cat. No. 324895) analog that is reported to be more potent and soluble than Salubrinal. In addition to being a useful tool in studying elF2α-mediated cellular processes in cultures in vitro, enhanced elF2α phosphorylation by Sal003 treatment in the brain (40 pmol per animal) is shown to impair long-term memory in mice in vivo.
A cell-permeable Salubrinal (Cat. No. 324895) analog that is reported to be more potent and soluble than Salubrinal. In addition to being a useful tool in studying elF2α-mediated cellular processes in cultures in vitro, enhanced elF2α phosphorylation by Sal003 treatment in the brain (40 pmol per animal) is shown to impair long-term memory in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Raven, J.F., et al. 2008. J. Biol. Chem.283, 3097.
Baltzis, D., et al. 2007. J. Biol. Chem.282, 31675.
Kazemi, S., et al. 2007. Mol. Biol. Cell18, 3635.
Costa-Mattioli, M., et al. 2007. Cell129, 195.
Robert, F., et al. 2006. Mol. Biol. Cell17, 4632.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

324896-1.1ML:
324896-MG:
324896-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service