Skip to Content
Merck
All Photos(1)

Documents

Safety Information

219673-M

Millipore

CFTR-F508del Corrector II, Corr-4a

A cell-permeable bisaminomethylbithiazole compound that is reported to rescue/correct the folding defect of CFTR δF508 mutant via direct interaction.

Synonym(s):

CFTR-F508del Corrector II, Corr-4a, Cystic Fibrosis Transmembrane Conductor Regulator Mutant Corrector II, N-(2-(5-Chloro-2-methoxy-phenylamino)-4ʹ-methyl-[4,5ʹ]bithiazolyl-2ʹ-yl)-benzamide, Corr-4, N-(2-(5-Chloro-2-methoxy-phenylamino)-4ʹ-methyl-[4,5ʹ]bithiazolyl-2ʹ-yl)-benzamide, Corr-4, Cystic Fibrosis Transmembrane Conductor Regulator Mutant Corrector II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17ClN4O2S2
CAS Number:
Molecular Weight:
456.97
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C21H17ClN4O2S2/c1-12-18(30-21(23-12)26-19(27)13-6-4-3-5-7-13)16-11-29-20(25-16)24-15-10-14(22)8-9-17(15)28-2/h3-11H,1-2H3,(H,24,25)(H,23,26,27)

InChI key

RDOBOPJBMQURAT-UHFFFAOYSA-N

General description

A cell-permeable bisaminomethylbithiazole compound that is reported to rescue/correct the folding defect of CFTR ΔF508 mutant via direct interaction and promote proper trafficking/glycosylation and surface expression in both transfected cells and primary airway epithelial cells from ΔF508-homozygous patients, while exhibiting little correcting effect toward N1303K-CFTR, P574H-CFTR, M345T-DRD4 (dopamine receptor 4), or G268V-ABCA1/CD243 /MDR1/ P-gp mutant. Reported to enhance the folding efficiency of both ΔF508 (by 3-fold at 10µM in FRT cells) and wild-type CFTR (by up to 70%), delay the ER-associated degradation of the core-glycosylated ΔF508 (by 30% at 10µM in FRT cells), and increase the stability of low-temperature resuced ΔF508 mutant on the plasma membrane (by 1.7-fold at 10µM in FRT cells). Corr-4a is shown to be more effective than VRT-325/CFcor-325 n restoring forskolin-stimulated CFTR activity in FRT and A549 ΔF508-CFTR transfectants. Unlike VRT-532 Corr-4a does not affect mutant CFTR channel gating activity.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Pedemonte, N., et al. 2010. Am. J. Physiol. Cell Physiol.298, C866.
Young, A., et al. 2009. Biochem. J.421, 377.
Wang, Y., et al. 2007. J. Biol. Chem.282, 33247.
Wang, Y., et al. 2007. Mol. Pharmacol.71, 751.
Wang, Y., et al. 2007. Biochem. J.406, 257.
Pedemonte, N., et al. 2005. J. Clin. Invest.115, 2564.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

219673-MG:
219673-10MG:
219673-10MG-M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service